Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029884.1 | NP_001025055.1 | 11 | 1547 | ||
XM_005267064.3 | XP_005267121.1 | 10 | 3626 | |||
XM_005267065.3 | XP_005267122.1 | 12 | 1573 | |||
XM_005267066.2 | XP_005267123.1 | 10 | 2430 | |||
XM_006715521.1 | XP_006715584.1 | 12 | 1800 | |||
XM_006715522.3 | XP_006715585.1 | 10 | 3667 | |||
XM_011535981.2 | XP_011534283.1 | 10 | 4423 | |||
XM_011535982.2 | XP_011534284.1 | 10 | 4423 | |||
XM_011535983.2 | XP_011534285.1 | 11 | 2848 | |||
XM_017011102.1 | XP_016866591.1 | 10 | 2432 | |||
GenBank mRNA | AB033035.1 | - | 4 | 420 | ||
AJ420468.1 | - | 8 | 785 | |||
AK056300.1 | - | 10 | 1446 | |||
AK293288.1 | - | 10 | 1478 | |||
AK294798.1 | - | 10 | 1601 | |||
AK307631.1 | - | 10 | 1500 | |||
BC089428.1 | - | 6 | 658 | |||
BC140864.1 | - | 9 | 1284 | 9021732 | ||
BC151134.1 | - | 9 | 1284 | 9057034 |