Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020726.4 | NP_065777.1 | 8 | 1477 | ||
XM_005248559.2 | XP_005248616.1 | 8 | 1468 | |||
XM_006714661.2 | XP_006714724.1 | 9 | 1532 | |||
XM_017009673.1 | XP_016865162.1 | 9 | 1532 | |||
GenBank mRNA | AB033052.2 | - | 8 | 1299 | ||
AJ300837.1 | - | 8 | 1424 | |||
AK027602.1 | - | 2 | 483 | |||
AK096416.1 | - | 8 | 1422 | |||
AK292803.1 | - | 8 | 1418 | |||
AL834241.1 | - | 1 | 184 | |||
BC001644.2 | - | 8 | 1415 | 2958628 | ||
BC004985.1 | - | 8 | 1415 | 2958628 |