Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESYT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020728.2 | NP_065779.1 | 3 | 688 | ||
GenBank mRNA | AB033054.2 | - | 4 | 474 | ||
AK126214.1 | - | 3 | 333 | |||
AK308824.1 | - | 3 | 296 | |||
AL833233.1 | - | 3 | 297 | |||
AY368150.1 | - | 3 | 688 | |||
DQ993201.1 | - | 5 | 706 |