Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007022.2 | NP_001007023.2 | 6 | 836 | ||
NM_001184765.1 | NP_001171694.1 | 6 | 843 | |||
NM_001184766.1 | NP_001171695.1 | 6 | 843 | |||
NM_020729.2 | NP_065780.2 | 6 | 843 | |||
XM_005271054.3 | XP_005271111.1 | 6 | 841 | |||
XM_011541815.2 | XP_011540117.1 | 6 | 841 | |||
XM_011541819.2 | XP_011540121.1 | 6 | 841 | |||
XM_011541821.2 | XP_011540123.1 | 6 | 841 | |||
XM_011541823.2 | XP_011540125.1 | 6 | 841 | |||
XM_011541824.2 | XP_011540126.1 | 6 | 841 | |||
XM_017001876.1 | XP_016857365.1 | 6 | 633 | |||
XM_017001877.1 | XP_016857366.1 | 6 | 633 | |||
XM_017001878.1 | XP_016857367.1 | 6 | 841 | |||
XM_017001879.1 | XP_016857368.1 | 6 | 841 | |||
XM_017001880.1 | XP_016857369.1 | 6 | 841 | |||
XM_017001881.1 | XP_016857370.1 | 5 | 713 | |||
GenBank mRNA | AB033055.1 | - | 6 | 756 | ||
AK302792.1 | - | 6 | 815 | |||
AK303209.1 | - | 6 | 840 | |||
AK308351.1 | - | 3 | 366 | |||
BC009779.1 | - | 3 | 357 | |||
BC029420.1 | - | 6 | 561 | |||
BC047368.1 | - | 6 | 783 | 5276959 | ||
BC091490.1 | - | 6 | 742 | 30367572 | ||
BI768058.1 | - | 5 | 665 | |||
BX499763.1 | - | 2 | 57 |