Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020733.1 | NP_065784.1 | 4 | 1292 | ||
XM_005247666.1 | XP_005247723.1 | 4 | 1287 | |||
GenBank mRNA | AB033063.2 | - | 3 | 887 |