Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282755.1 | NP_001269684.1 | 11 | 1409 | ||
NM_001282756.1 | NP_001269685.1 | 10 | 1061 | |||
NM_020744.3 | NP_065795.1 | 10 | 1136 | |||
XM_005264456.2 | XP_005264513.1 | 10 | 938 | |||
XM_005264458.2 | XP_005264515.1 | 10 | 938 | |||
XM_005264459.2 | XP_005264516.1 | 11 | 1409 | |||
XM_017004561.1 | XP_016860050.1 | 9 | 749 | |||
XM_017004562.1 | XP_016860051.1 | 10 | 938 | |||
GenBank mRNA | AB033092.1 | - | 10 | 1061 | ||
AK027304.1 | - | 10 | 934 | |||
AK127245.1 | - | 11 | 1409 | |||
BC004227.2 | - | 10 | 964 | 3542706 | ||
BC053631.1 | - | 10 | 1122 | 6472900 |