Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206491.1 | NP_001193420.1 | 6 | 2012 | ||
NM_020746.4 | NP_065797.2 | 7 | 2187 | |||
NR_037921.1 | 6 | 2014 | ||||
GenBank mRNA | AB033097.1 | - | 6 | 2093 | ||
AB232371.1 | - | 6 | 2087 | |||
AK023799.1 | - | 7 | 2142 | |||
AL833736.1 | - | 1 | 2242 | |||
BC044952.1 | - | 7 | 2121 | 5751684 | ||
BX649084.1 | - | 6 | 1973 |