Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF608 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020747.2 | NP_065798.2 | 5 | 3977 | ||
XM_005272037.2 | XP_005272094.1 | 6 | 4739 | |||
XM_005272038.1 | XP_005272095.1 | 6 | 4329 | |||
XM_005272039.2 | XP_005272096.1 | 6 | 4739 | |||
XM_005272040.3 | XP_005272097.1 | 5 | 3225 | |||
XM_011543520.2 | XP_011541822.1 | 6 | 4739 | |||
XM_011543522.2 | XP_011541824.1 | 5 | 3395 | |||
XM_011543523.2 | XP_011541825.1 | 4 | 2837 | |||
XM_017009674.1 | XP_016865163.1 | 6 | 4739 | |||
GenBank mRNA | AB033107.2 | - | 5 | 3978 | ||
BC038370.1 | - | 1 | 39 | |||
BC131507.1 | - | 4 | 1491 | |||
BC151226.1 | - | 5 | 3978 | 9007221 | ||
CR749624.1 | - | 5 | 3058 |