Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282944.1 | NP_001269873.1 | 3 | 394 | ||
NM_001282946.1 | NP_001269875.1 | 3 | 394 | |||
NM_015160.2 | NP_055975.1 | 3 | 394 | |||
XM_005266059.4 | XP_005266116.1 | 3 | 309 | |||
XM_017014543.1 | XP_016870032.1 | 2 | 227 | |||
GenBank mRNA | AK296617.1 | - | 3 | 306 | ||
AK299304.1 | - | 3 | 326 | |||
AK307206.1 | - | 3 | 307 | |||
AK308104.1 | - | 3 | 309 | |||
BC022949.1 | - | 3 | 286 | |||
BC033103.1 | - | 3 | 303 | |||
BC111399.1 | - | 3 | 309 | 4609491 | ||
BC132724.1 | - | 3 | 333 | 40146746 | ||
BC136599.1 | - | 3 | 333 | 9020589 | ||
D21064.1 | - | 3 | 307 | |||
D50913.2 | - | 3 | 307 |