Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020759.2 | NP_065810.2 | 23 | 6845 | ||
XM_011521831.2 | XP_011520133.1 | 23 | 6858 | |||
XM_011521832.2 | XP_011520134.1 | 23 | 6855 | |||
XM_011521833.2 | XP_011520135.1 | 22 | 6825 | |||
XM_011521834.2 | XP_011520136.1 | 22 | 6795 | |||
XM_011521835.2 | XP_011520137.1 | 21 | 6717 | |||
XM_011521836.2 | XP_011520138.1 | 23 | 7160 | |||
XM_011521837.2 | XP_011520139.1 | 21 | 6627 | |||
XM_011521839.2 | XP_011520141.1 | 10 | 5627 | |||
XM_017022439.1 | XP_016877928.1 | 22 | 6780 | |||
XM_017022440.1 | XP_016877929.1 | 22 | 6757 | |||
XM_017022441.1 | XP_016877930.1 | 21 | 7076 | |||
XM_017022442.1 | XP_016877931.1 | 16 | 6184 | |||
XM_017022443.1 | XP_016877932.1 | 15 | 6168 | |||
GenBank mRNA | CR749416.1 | - | 1 | 867 |