Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163034.1 | NP_001156506.1 | 5 | 1332 | ||
NM_020761.2 | NP_065812.1 | 5 | 1332 | |||
GenBank mRNA | AB082951.1 | - | 5 | 527 | ||
AK055912.1 | - | 5 | 1178 | |||
AK295964.1 | - | 4 | 1255 | |||
AK296321.1 | - | 5 | 1327 | |||
AY090663.1 | - | 5 | 889 | |||
BC064515.1 | - | 5 | 1280 | |||
BC136652.1 | - | 5 | 722 | 9020642 | ||
BC136654.1 | - | 5 | 722 | 9020644 |