Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163034.1 | NP_001156506.1 | 12 | 2158 | ||
NM_020761.2 | NP_065812.1 | 12 | 2158 | |||
GenBank mRNA | AB037724.1 | - | 8 | 840 | ||
AB082951.1 | - | 12 | 1353 | |||
AK055912.1 | - | 12 | 2004 | |||
AK295964.1 | - | 11 | 2081 | |||
AK296321.1 | - | 12 | 2153 | |||
AY090663.1 | - | 12 | 1715 | |||
BC136652.1 | - | 12 | 1548 | 9020642 | ||
BC136654.1 | - | 12 | 1548 | 9020644 |