Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020762.2 | NP_065813.1 | 19, 20 | 2464 | ||
XM_005269042.3 | XP_005269099.1 | 19, 20 | 2784 | |||
XM_011538580.1 | XP_011536882.1 | 20, 21 | 2572 | |||
XM_011538581.2 | XP_011536883.1 | 20, 21 | 4077 | |||
GenBank mRNA | AB037725.1 | - | 18, 19 | 3686 | ||
AK023899.1 | - | 9, 10 | 1194 | |||
BC053903.1 | - | 19, 20 | 2464 | 6526787 |