Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105243.1 | NP_001098713.1 | 2 | 4081 | ||
NM_001184880.1 | NP_001171809.1 | 3 | 4222 | |||
NM_020766.2 | NP_065817.2 | 2 | 4081 | |||
XM_011530997.2 | XP_011529299.1 | 3 | 3191 | |||
GenBank mRNA | AB037734.1 | - | 2 | 296 | ||
AK096591.1 | - | 2 | 1034 | |||
BC136628.1 | - | 2 | 962 | 9020618 | ||
CR749278.1 | - | 2 | 962 | |||
EF676096.1 | - | 3 | 4222 |