Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020778.4 | NP_065829.3 | 4 | 1110 | ||
XM_017022444.1 | XP_016877933.1 | 3 | 1108 | |||
XM_017022445.1 | XP_016877934.1 | 4 | 1159 | |||
GenBank mRNA | AY044449.1 | - | 4 | 1110 |