Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006657.1 | NP_001006658.1 | 28 | 3574 | ||
NM_020779.3 | NP_065830.2 | 27 | 3541 | |||
XM_011533007.2 | XP_011531309.1 | 17 | 2266 | |||
GenBank mRNA | AB037757.2 | - | 28 | 3582 | ||
AK091205.1 | - | 18 | 2392 | |||
AK122917.1 | - | 27 | 3466 | |||
AK315094.1 | - | 27 | 3370 | |||
BC036659.2 | - | 27 | 3540 | 5266940 |