Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 24 | 3182 | ||
XM_005248177.1 | XP_005248234.1 | 23 | 3137 | |||
XM_011513872.2 | XP_011512174.1 | 23 | 3137 | |||
XM_017008482.1 | XP_016863971.1 | 21 | 2931 | |||
GenBank mRNA | AB037766.1 | - | 21 | 2920 | ||
AK025543.1 | - | 9 | 1267 | |||
BX647334.1 | - | 23 | 3107 | |||
EU450799.1 | - | 24 | 3009 |