Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080522.2 | NP_001073991.2 | 19 | 2488 | ||
XM_005248177.1 | XP_005248234.1 | 18 | 2443 | |||
XM_011513872.2 | XP_011512174.1 | 18 | 2443 | |||
XM_017008482.1 | XP_016863971.1 | 16 | 2237 | |||
GenBank mRNA | AB037766.1 | - | 16 | 2226 | ||
AK023876.1 | - | 10 | 1398 | |||
AK025543.1 | - | 4 | 573 | |||
BX647334.1 | - | 18 | 2413 | |||
EU450799.1 | - | 19 | 2315 |