Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001851.4 | NP_001842.3 | 19 | 1535 | ||
NM_078485.3 | NP_511040.2 | 13 | 803 | |||
XM_011535429.2 | XP_011533731.1 | 19 | 1535 | |||
XM_011535430.2 | XP_011533732.1 | 13 | 801 | |||
XM_017010246.1 | XP_016865735.1 | 16 | 892 | |||
XM_017010247.1 | XP_016865736.1 | 10 | 748 | |||
GenBank mRNA | AK125738.1 | - | 9 | 468 | ||
BC008620.1 | - | 13 | 730 | |||
BC063646.1 | - | 13 | 777 | 4914907 | ||
X54412.1 | - | 19 | 1507 |