Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292003.1 | NP_001278932.1 | 7 | 1643 | ||
NM_001292006.1 | NP_001278935.1 | 7 | 1372 | |||
NM_001292007.1 | NP_001278936.1 | 6 | 1051 | |||
NM_020803.4 | NP_065854.3 | 7 | 1600 | |||
NR_120447.1 | 5 | 864 | ||||
GenBank mRNA | AB037799.2 | - | 6 | 1283 | ||
AK128827.1 | - | 7 | 1643 | |||
BC041384.2 | - | 7 | 1550 | 5275946 | ||
BC041901.1 | - | 5 | 720 | |||
BX538173.1 | - | 7 | 1517 | |||
BX640727.1 | - | 7 | 1510 | |||
BX640744.1 | - | 7 | 1301 | |||
BX647454.1 | - | 7 | 1535 | |||
BX648367.1 | - | 5 | 568 | |||
CR749265.1 | - | 6 | 969 |