Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258415.1 | NP_001245344.1 | 6, 7 | 913 | ||
NM_001258416.1 | NP_001245345.1 | 7, 8 | 799 | |||
NM_001258417.1 | NP_001245346.1 | 6, 7 | 613 | |||
NM_001258418.1 | NP_001245347.1 | 6, 7 | 731 | |||
NM_020809.3 | NP_065860.2 | 7, 8 | 910 | |||
XM_006718893.3 | XP_006718956.1 | 6, 7 | 819 | |||
GenBank mRNA | AB037812.1 | - | 6, 7 | 635 | ||
AK307602.1 | - | 6, 7 | 819 | |||
AY496263.1 | - | 7, 8 | 910 | |||
AY496264.1 | - | 6, 7 | 613 | |||
AY496265.1 | - | 6, 7 | 731 | |||
AY496266.1 | - | 6, 7 | 913 | |||
AY496267.1 | - | 7, 8 | 799 |