Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL9A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001852.3 | NP_001843.1 | 11 | 609 | ||
XM_006710365.3 | XP_006710428.1 | 12 | 717 | |||
XM_011540715.2 | XP_011539017.1 | 11 | 1197 | |||
XM_011540716.2 | XP_011539018.1 | 10 | 677 | |||
XM_011540717.2 | XP_011539019.1 | 10 | 1525 | |||
XM_017000332.1 | XP_016855821.1 | 11 | 663 | |||
XM_017000333.1 | XP_016855822.1 | 10 | 663 | |||
GenBank mRNA | BC041479.1 | - | 10 | 842 | ||
BC136326.1 | - | 11 | 602 | 9020313 | ||
BC136327.1 | - | 11 | 602 | 9020314 | ||
M95610.1 | - | 11 | 498 |