Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321768.1 | NP_001308697.1 | 4 | 357 | ||
NM_020813.3 | NP_065864.2 | 5 | 484 | |||
XM_011527148.1 | XP_011525450.1 | 5 | 3311 | |||
XM_017027032.1 | XP_016882521.1 | 4 | 3184 | |||
GenBank mRNA | AB037817.1 | - | 2 | 127 | ||
AF352026.1 | - | 5 | 468 | |||
AK291416.1 | - | 5 | 476 | |||
AL831845.1 | - | 5 | 484 | |||
BC125221.1 | - | 5 | 441 | 40124078 | ||
BC125222.1 | - | 5 | 443 | 40124080 |