Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020814.2 | NP_065865.1 | 4 | 2720 | ||
GenBank mRNA | AB037820.1 | - | 4 | 2720 | ||
BC048793.1 | - | 4 | 794 | |||
BC098448.1 | - | 4 | 1149 | 6209260 |