Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020817.1 | NP_065868.1 | 5, 6 | 749 | ||
XM_006713715.1 | XP_006713778.1 | 4, 5 | 691 | |||
XM_011513051.1 | XP_011511353.1 | 4, 5 | 1198 | |||
XM_011513052.2 | XP_011511354.1 | 3, 4 | 505 | |||
XM_011513053.1 | XP_011511355.1 | 4, 5 | 641 | |||
XM_011513054.2 | XP_011511356.1 | 4, 5 | 477 | |||
XM_017006956.1 | XP_016862445.1 | 3, 4 | 611 | |||
XM_017006957.1 | XP_016862446.1 | 4, 5 | 1296 | |||
XM_017006958.1 | XP_016862447.1 | 4, 5 | 1143 | |||
XM_017006959.1 | XP_016862448.1 | 2, 3 | 296 | |||
GenBank mRNA | AB037828.1 | - | 1, 2 | 27 | ||
AF509494.1 | - | 5, 6 | 749 | |||
AK125304.1 | - | 5, 6 | 653 | |||
AK295855.1 | - | 3, 4 | 494 | |||
AK302488.1 | - | 5, 6 | 750 | |||
BC150592.1 | - | 4, 5 | 691 |