Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020817.1 | NP_065868.1 | 10 | 1935 | ||
XM_005247681.3 | XP_005247738.1 | 4 | 920 | |||
XM_006713715.1 | XP_006713778.1 | 9 | 1877 | |||
XM_011513051.1 | XP_011511353.1 | 9 | 2384 | |||
XM_011513052.2 | XP_011511354.1 | 8 | 1691 | |||
XM_011513053.1 | XP_011511355.1 | 9 | 1827 | |||
XM_011513054.2 | XP_011511356.1 | 9 | 1663 | |||
XM_017006956.1 | XP_016862445.1 | 8 | 1797 | |||
XM_017006957.1 | XP_016862446.1 | 9 | 2482 | |||
XM_017006958.1 | XP_016862447.1 | 9 | 2329 | |||
XM_017006959.1 | XP_016862448.1 | 7 | 1423 | |||
GenBank mRNA | AB037828.1 | - | 6 | 1213 | ||
AF509494.1 | - | 10 | 1935 | |||
AK125304.1 | - | 10 | 1839 | |||
AK295855.1 | - | 8 | 1680 | |||
AK302488.1 | - | 10 | 1936 | |||
BC150592.1 | - | 9 | 1877 |