Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PREX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020820.3 | NP_065871.2 | 8, 9 | 1061 | ||
GenBank mRNA | AB037836.2 | - | 8, 9 | 924 | ||
AJ320261.1 | - | 8, 9 | 1061 | |||
AL832913.1 | - | 1, 2 | 30 | |||
BC053616.1 | - | 4, 5 | 511 |