Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020824.3 | NP_065875.3 | 9 | 2205 | ||
XM_005252542.3 | XP_005252599.1 | 9 | 2273 | |||
XM_005252544.4 | XP_005252601.1 | 8 | 3928 | |||
XM_011519602.2 | XP_011517904.1 | 9 | 3958 | |||
XM_011519603.2 | XP_011517905.1 | 9 | 2365 | |||
XM_011519604.2 | XP_011517906.1 | 9 | 2181 | |||
XM_011519605.2 | XP_011517907.1 | 8 | 3928 | |||
XM_011519606.2 | XP_011517908.1 | 6 | 3823 | |||
XM_011519607.2 | XP_011517909.1 | 9 | 3958 | |||
XM_017016459.1 | XP_016871948.1 | 9 | 2365 | |||
XM_017016460.1 | XP_016871949.1 | 8 | 2335 | |||
XM_017016461.1 | XP_016871950.1 | 8 | 2146 | |||
GenBank mRNA | AB037845.2 | - | 8 | 1678 | ||
AF480466.1 | - | 9 | 2152 | |||
AK090642.1 | - | 1 | 420 | |||
AK295649.1 | - | 2 | 1295 | |||
AK297278.1 | - | 3 | 1321 | |||
AK307604.1 | - | 9 | 2182 | |||
BC118915.2 | - | 7 | 1779 | 40071194 | ||
BX537570.1 | - | 1 | 1207 |