Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRAMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020825.3 | NP_065876.3 | 20 | 3672 | ||
GenBank mRNA | AB037847.2 | - | 18 | 3065 | ||
AK095591.1 | - | 1 | 1011 | |||
BC137172.1 | - | 19 | 3222 | 9021169 | ||
BC137173.1 | - | 19 | 3222 | 9021170 | ||
BC152477.1 | - | 18 | 3065 | 8862615 |