Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292033.1 | NP_001278962.1 | 2 | 1083 | ||
NM_020827.2 | NP_065878.1 | 2 | 970 | |||
XM_017008483.1 | XP_016863972.1 | 2 | 1228 | |||
XM_017008484.1 | XP_016863973.1 | 2 | 862 | |||
XM_017008485.1 | XP_016863974.1 | 4 | 1095 | |||
XM_017008486.1 | XP_016863975.1 | 2 | 862 | |||
GenBank mRNA | AB037851.1 | - | 1 | 810 | ||
AK091993.1 | - | 2 | 901 | |||
BC030535.1 | - | 2 | 1082 | 5240222 |