Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304763.1 | NP_001291692.1 | 3 | 1032 | ||
NM_001304764.1 | NP_001291693.1 | 2 | 233 | |||
NM_020832.2 | NP_065883.1 | 2 | 163 | |||
XM_005245366.3 | XP_005245423.1 | 2 | 601 | |||
XM_011509811.2 | XP_011508113.1 | 2 | 210 | |||
XM_011509812.2 | XP_011508114.1 | 3 | 480 | |||
XM_011509813.2 | XP_011508115.1 | 2 | 364 | |||
GenBank mRNA | AB037862.1 | - | 3 | 1032 | ||
BC032463.1 | - | 2 | 160 | 5196247 | ||
CR749307.1 | - | 2 | 233 | |||
DQ655935.2 | - | 2 | 662 |