Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320239.1 | NP_001307168.1 | 2 | 441 | ||
NM_001320240.1 | NP_001307169.1 | 2 | 441 | |||
NM_020850.2 | NP_065901.1 | 2 | 441 | |||
XM_005256070.2 | XP_005256127.1 | 2 | 349 | |||
XM_005256072.2 | XP_005256129.1 | 2 | 349 | |||
XM_011523245.2 | XP_011521547.1 | 2 | 349 | |||
XM_017023508.1 | XP_016878997.1 | 2 | 349 | |||
GenBank mRNA | AB040897.1 | - | 2 | 330 | ||
AK055727.1 | - | 2 | 346 | |||
AK295171.1 | - | 2 | 349 | |||
AK298806.1 | - | 2 | 368 | |||
AK304030.1 | - | 2 | 346 | |||
AY337313.1 | - | 2 | 441 | |||
BC099917.1 | - | 2 | 346 | 30528481 | ||
BC121176.1 | - | 2 | 439 | 40122561 | ||
BC121177.2 | - | 2 | 439 | 40122562 |