Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL11A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080679.2 | NP_542410.2 | 61 | 4713 | ||
NM_080680.2 | NP_542411.2 | 64 | 5034 | |||
NM_080681.2 | NP_542412.2 | 62 | 4776 | |||
XM_011514299.2 | XP_011512601.1 | 59 | 4325 | |||
XM_011514300.2 | XP_011512602.1 | 58 | 4145 | |||
XM_011514302.2 | XP_011512604.1 | 57 | 4346 | |||
XM_017010250.1 | XP_016865739.1 | 65 | 5299 | |||
XM_017010251.1 | XP_016865740.1 | 57 | 4163 | |||
GenBank mRNA | AK130938.1 | - | 7 | 597 | ||
J04974.1 | - | 37 | 2619 |