Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029939.3 | NP_001025110.2 | 6 | 1033 | ||
NM_001167583.2 | NP_001161055.1 | 5 | 874 | |||
NM_001174157.1 | NP_001167628.1 | 4 | 765 | |||
NM_001174158.1 | NP_001167629.1 | 5 | 874 | |||
NM_001289394.1 | NP_001276323.1 | 6 | 1110 | |||
NM_020863.3 | NP_065914.2 | 5 | 951 | |||
NR_110323.1 | 5 | 951 | ||||
XM_011517203.1 | XP_011515505.1 | 5 | 848 | |||
XM_011517204.2 | XP_011515506.1 | 4 | 671 | |||
XM_011517206.1 | XP_011515508.1 | 5 | 872 | |||
XM_017013716.1 | XP_016869205.1 | 4 | 746 | |||
GenBank mRNA | AB040918.1 | - | 3 | 349 | ||
AB167738.1 | - | 5 | 940 | |||
AB167739.1 | - | 6 | 1099 | |||
AB167740.1 | - | 6 | 1026 | |||
AB167741.1 | - | 5 | 867 | |||
AK094089.1 | - | 4 | 748 | |||
AK300133.1 | - | 5 | 863 | |||
AK300324.1 | - | 5 | 870 | |||
AK300471.1 | - | 5 | 936 | |||
AK311494.1 | - | 5 | 872 | |||
AL832066.1 | - | 5 | 951 | |||
BC012989.2 | - | 5 | 865 | |||
BC025423.2 | - | 3 | 543 | |||
BC098405.1 | - | 5 | 874 | |||
BC101766.1 | - | 5 | 864 | 8069272 | ||
BC101768.1 | - | 5 | 864 | 8069274 | ||
BC143519.1 | - | 4 | 679 | 9052028 |