Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020866.2 | NP_065917.1 | 2 | 1447 | ||
XM_017020678.1 | XP_016876167.1 | 2 | 367 | |||
XM_017020679.1 | XP_016876168.1 | 1 | 60 | |||
GenBank mRNA | AB040923.1 | - | 2 | 1413 | ||
AF252283.1 | - | 2 | 1281 | |||
AK054609.1 | - | 2 | 659 | |||
AK291435.1 | - | 2 | 1447 | |||
BC022460.1 | - | 2 | 691 | 4819077 |