Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286725.1 | NP_001273654.1 | 6 | 2165 | ||
NM_020866.2 | NP_065917.1 | 7 | 2348 | |||
XM_017020678.1 | XP_016876167.1 | 7 | 1268 | |||
XM_017020679.1 | XP_016876168.1 | 7 | 1855 | |||
GenBank mRNA | AB040923.1 | - | 7 | 2314 | ||
AF252283.1 | - | 7 | 2182 | |||
AK054609.1 | - | 7 | 1560 | |||
AK291435.1 | - | 7 | 2348 | |||
AK295939.1 | - | 6 | 1356 | |||
BC022460.1 | - | 7 | 1592 | 4819077 |