Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020870.3 | NP_065921.2 | 3 | 833 | ||
GenBank mRNA | AK021429.1 | - | 3 | 694 | ||
BC033203.1 | - | 2 | 379 | |||
BC041023.1 | - | 1 | 83 | |||
BC053671.1 | - | 671 | 6144754 |