Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199417.1 | NP_001186346.1 | 5, 6 | 438 | ||
XM_006721991.1 | XP_006722054.1 | 5, 6 | 479 | |||
XM_006721992.3 | XP_006722055.1 | 5, 6 | 466 | |||
XM_006721993.3 | XP_006722056.1 | 5, 6 | 465 | |||
XM_006721994.3 | XP_006722057.1 | 5, 6 | 464 | |||
XM_011525073.1 | XP_011523375.1 | 5, 6 | 398 | |||
XM_011525074.2 | XP_011523376.1 | 5, 6 | 497 | |||
XM_011525076.2 | XP_011523378.1 | 5, 6 | 464 | |||
XM_011525077.2 | XP_011523379.1 | 5, 6 | 464 | |||
XM_017024902.1 | XP_016880391.1 | 5, 6 | 464 | |||
GenBank mRNA | EG328307.1 | - | 2, 3 | 171 |