Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 18, 19 | 3624 | ||
NM_020902.1 | NP_065953.1 | 16, 17 | 3543 | |||
XM_005272491.3 | XP_005272548.1 | 17, 18 | 3560 | |||
XM_011528154.2 | XP_011526456.1 | 18, 19 | 3543 | |||
GenBank mRNA | AB040976.1 | - | 9, 10 | 2446 | ||
AF289580.1 | - | 6, 7 | 1496 | |||
AL833927.1 | - | 6, 7 | 2226 | |||
BC020431.1 | - | 18, 19 | 3467 | |||
BC128201.1 | - | 2, 3 | 142 | |||
BC128202.1 | - | 2, 3 | 142 |