Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080429.2 | NP_001073898.1 | 3 | 554 | ||
NM_020902.1 | NP_065953.1 | 3 | 554 | |||
XM_005272491.3 | XP_005272548.1 | 3 | 523 | |||
XM_011528154.2 | XP_011526456.1 | 3 | 524 | |||
XM_017027036.1 | XP_016882525.1 | 3 | 527 | |||
GenBank mRNA | BC020431.1 | - | 3 | 397 |