Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161354.1 | NP_001154826.1 | 9 | 1547 | ||
NM_020904.2 | NP_065955.2 | 10 | 1622 | |||
XM_005259107.2 | XP_005259164.1 | 9 | 1542 | |||
XM_006723301.2 | XP_006723364.1 | 9 | 1542 | |||
XM_011527157.1 | XP_011525459.1 | 10 | 1629 | |||
XM_011527158.2 | XP_011525460.1 | 9 | 1468 | |||
XM_011527159.1 | XP_011525461.1 | 10 | 1642 | |||
XM_011527160.1 | XP_011525462.1 | 10 | 1629 | |||
XM_011527162.2 | XP_011525464.1 | 3 | 238 | |||
XM_017027037.1 | XP_016882526.1 | 7 | 906 | |||
GenBank mRNA | AB208908.1 | - | 10 | 1584 | ||
AY007233.1 | - | 10 | 1596 | |||
BC024157.2 | - | 10 | 1602 | 3953478 | ||
BC033832.1 | - | 2 | 72 | |||
BC064601.1 | - | 9 | 1374 | 5785991 |