Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297619.1 | NP_001284548.1 | 5 | 620 | ||
NM_020917.2 | NP_065968.1 | 5 | 617 | |||
XM_017027038.1 | XP_016882527.1 | 5 | 897 | |||
GenBank mRNA | AB046779.1 | - | 4 | 590 | ||
BC144691.1 | - | 4 | 590 | |||
BC152446.1 | - | 4 | 590 | 8862575 |