Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL16A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001856.3 | NP_001847.3 | 68 | 4704 | ||
XM_005270481.1 | XP_005270538.1 | 67 | 4651 | |||
XM_011540722.1 | XP_011539024.1 | 67 | 4654 | |||
XM_011540723.1 | XP_011539025.1 | 66 | 4606 | |||
XM_011540724.1 | XP_011539026.1 | 67 | 4576 | |||
XM_011540726.2 | XP_011539028.1 | 63 | 4122 | |||
XM_011540727.2 | XP_011539029.1 | 58 | 3460 | |||
XM_011540728.2 | XP_011539030.1 | 46 | 2942 | |||
XM_017000338.1 | XP_016855827.1 | 67 | 4666 | |||
XM_017000339.1 | XP_016855828.1 | 67 | 4663 | |||
XM_017000340.1 | XP_016855829.1 | 45 | 2905 | |||
XM_017000341.1 | XP_016855830.1 | 45 | 2695 | |||
GenBank mRNA | AB209571.1 | - | 62 | 3918 | ||
AF370368.1 | - | 12 | 1888 | |||
M92642.1 | - | 69 | 4489 | |||
S57132.1 | - | 51 | 3085 |