Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293274.1 | NP_001280203.1 | 3, 4 | 792 | ||
NM_020925.2 | NP_065976.2 | 3, 4 | 368 | |||
XM_011541863.1 | XP_011540165.1 | 3, 4 | 792 | |||
GenBank mRNA | AB046793.1 | - | 1, 2 | 150 | ||
BC039301.1 | - | 2, 3 | 323 | 4836806 | ||
BX537603.1 | - | 3, 4 | 791 | |||
CX163220.1 | - | 3, 4 | 359 |