Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190791.1 | NP_001177720.1 | 6 | 1447 | ||
NM_020933.4 | NP_065984.3 | 7 | 1543 | |||
NR_102435.1 | 7 | 1689 | ||||
GenBank mRNA | AB046808.1 | - | 6 | 1320 | ||
AF148135.1 | - | 6 | 1430 | |||
AF275255.1 | - | 7 | 1526 | |||
AF307096.1 | - | 4 | 1664 | |||
AF307097.1 | - | 5 | 1762 | |||
AK056205.1 | - | 1 | 407 | |||
AK056570.1 | - | 7 | 1526 | |||
AK314560.1 | - | 7 | 1526 | |||
AL834126.2 | - | 5 | 2618 | |||
BC078154.1 | - | 7 | 1513 | 6060418 |