Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020936.2 | NP_065987.1 | 12 | 1382 | ||
NR_135104.1 | 12 | 1454 | ||||
NR_135105.1 | 12 | 1382 | ||||
XM_017019710.1 | XP_016875199.1 | 12 | 1421 | |||
XM_017019711.1 | XP_016875200.1 | 11 | 1476 | |||
XM_017019712.1 | XP_016875201.1 | 12 | 1274 | |||
XM_017019713.1 | XP_016875202.1 | 12 | 1397 | |||
XM_017019714.1 | XP_016875203.1 | 13 | 1555 | |||
XM_017019717.1 | XP_016875206.1 | 12 | 1319 | |||
XM_017019718.1 | XP_016875207.1 | 10 | 1737 | |||
XM_017019719.1 | XP_016875208.1 | 12 | 1274 | |||
XM_017019720.1 | XP_016875209.1 | 10 | 2352 | |||
GenBank mRNA | AB046815.1 | - | 7 | 3209 | ||
AK301043.1 | - | 12 | 1417 | |||
AK307384.1 | - | 12 | 1358 | |||
AL833756.1 | - | 10 | 2419 | |||
BC030020.2 | - | 12 | 1326 | 4821394 | ||
BC035911.1 | - | 1 | 231 |