Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308133.1 | NP_001295062.1 | 9, 10 | 1806 | ||
NM_001308134.1 | NP_001295063.1 | 10, 11 | 1884 | |||
NM_020937.3 | NP_065988.1 | 10, 11 | 1884 | |||
XM_011537034.2 | XP_011535336.1 | 10, 11 | 1883 | |||
XM_011537035.2 | XP_011535337.1 | 9, 10 | 1805 | |||
XM_017021523.1 | XP_016877012.1 | 10, 11 | 1883 | |||
XM_017021524.1 | XP_016877013.1 | 8, 9 | 1105 | |||
XM_017021525.1 | XP_016877014.1 | 8, 9 | 1672 | |||
XM_017021526.1 | XP_016877015.1 | 7, 8 | 916 | |||
XM_017021527.1 | XP_016877016.1 | 9, 10 | 1697 | |||
GenBank mRNA | AK093422.1 | - | 10, 11 | 1883 | ||
BC036056.1 | - | 10, 11 | 1885 | |||
BC140776.1 | - | 9, 10 | 1791 | 9021644 | ||
BC144511.1 | - | 9, 10 | 1791 | 9053038 | ||
DQ140356.1 | - | 10, 11 | 1891 |