Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020944.2 | NP_065995.1 | 9 | 1968 | ||
XM_005251526.4 | XP_005251583.1 | 9 | 2013 | |||
XM_006716809.3 | XP_006716872.1 | 9 | 2013 | |||
XM_011517973.2 | XP_011516275.1 | 9 | 1573 | |||
XM_017014937.1 | XP_016870426.1 | 9 | 1995 | |||
XM_017014938.1 | XP_016870427.1 | 9 | 2013 | |||
XM_017014939.1 | XP_016870428.1 | 9 | 1573 | |||
XM_017014940.1 | XP_016870429.1 | 8 | 1354 | |||
XM_017014941.1 | XP_016870430.1 | 8 | 1776 | |||
XM_017014942.1 | XP_016870431.1 | 9 | 1586 | |||
XM_017014943.1 | XP_016870432.1 | 9 | 1568 | |||
XM_017014944.1 | XP_016870433.1 | 8 | 1031 | |||
XM_017014945.1 | XP_016870434.1 | 8 | 1013 | |||
XM_017014946.1 | XP_016870435.1 | 10 | 1624 | |||
GenBank mRNA | AB046825.1 | - | 9 | 1958 | ||
AF258662.1 | - | 6 | 1016 | |||
AJ309567.1 | - | 9 | 1968 | |||
AK295967.1 | - | 10 | 1961 | |||
AK297435.1 | - | 7 | 975 | |||
AL834306.1 | - | 9 | 1728 | |||
BC011363.1 | - | 9 | 1927 | 4339156 |