Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020944.2 | NP_065995.1 | 15 | 2767 | ||
XM_005251526.4 | XP_005251583.1 | 15 | 2746 | |||
XM_006716809.3 | XP_006716872.1 | 15 | 2812 | |||
XM_011517973.2 | XP_011516275.1 | 15 | 2372 | |||
XM_017014937.1 | XP_016870426.1 | 15 | 2728 | |||
XM_017014938.1 | XP_016870427.1 | 15 | 2812 | |||
XM_017014939.1 | XP_016870428.1 | 15 | 2306 | |||
XM_017014940.1 | XP_016870429.1 | 14 | 2153 | |||
XM_017014941.1 | XP_016870430.1 | 14 | 2575 | |||
XM_017014942.1 | XP_016870431.1 | 15 | 2385 | |||
XM_017014943.1 | XP_016870432.1 | 15 | 2367 | |||
XM_017014944.1 | XP_016870433.1 | 14 | 1830 | |||
XM_017014945.1 | XP_016870434.1 | 14 | 1812 | |||
XM_017014946.1 | XP_016870435.1 | 16 | 2423 | |||
GenBank mRNA | AB046825.1 | - | 15 | 2757 | ||
AF258662.1 | - | 12 | 1815 | |||
AJ309567.1 | - | 15 | 2767 | |||
AK027884.1 | - | 15 | 2430 | |||
AK057610.1 | - | 1 | 529 | |||
AK295967.1 | - | 16 | 2760 | |||
AK297435.1 | - | 13 | 1774 | |||
AL834306.1 | - | 15 | 2527 | |||
BC011363.1 | - | 15 | 2726 | 4339156 |