Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020946.1 | NP_065997.1 | 6 | 555 | ||
NM_024820.2 | NP_079096.2 | 6 | 555 | |||
XM_005252109.2 | XP_005252166.1 | 6 | 404 | |||
XM_005252111.4 | XP_005252168.1 | 6 | 1371 | |||
XM_005252113.4 | XP_005252170.1 | 6 | 1360 | |||
XM_006717195.3 | XP_006717258.1 | 6 | 1372 | |||
XM_011518882.2 | XP_011517184.1 | 6 | 1374 | |||
XM_011518883.2 | XP_011517185.1 | 6 | 1375 | |||
XM_011518884.2 | XP_011517186.1 | 5 | 1325 | |||
XM_011518885.2 | XP_011517187.1 | 5 | 1067 | |||
XM_011518886.2 | XP_011517188.1 | 6 | 1372 | |||
XM_011518887.2 | XP_011517189.1 | 6 | 1363 | |||
XM_017014947.1 | XP_016870436.1 | 5 | 1330 | |||
XM_017014948.1 | XP_016870437.1 | 6 | 404 | |||
XM_017014949.1 | XP_016870438.1 | 6 | 1375 | |||
XM_017014950.1 | XP_016870439.1 | 6 | 1374 | |||
XM_017014951.1 | XP_016870440.1 | 5 | 348 | |||
XM_017014952.1 | XP_016870441.1 | 5 | 531 | |||
GenBank mRNA | AK024782.1 | - | 6 | 543 | ||
AK225104.1 | - | 6 | 543 | |||
AK295710.1 | - | 4 | 416 | |||
BC009616.2 | - | 5 | 523 | |||
BC028061.1 | - | 5 | 345 | |||
BC039703.1 | - | 6 | 408 | 4999484 |